Human Recombinant G-CSFR/CD114/CSF3R (from HEK293 Cells)
Supplier: Sino Biological
10218-HCCH-100
103624-034EA 300.98 USD
103624-034
Human Recombinant G-CSFR/CD114/CSF3R (from HEK293 Cells)
Proteins and Peptides
The mature form of human G-CSF receptor (NP_000751.1) extracellular domain (Met 1-Pro 621) with five aminio acids at the C-terminus was expressed and purified.