Human Recombinant EG-VEGF/prokineticin-1 Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10183-H08B-20 10183-H08B-50 10183-H08B-50
103678-512EA 199.98 USD
103678-512 CA76454-086 103678-510
Human Recombinant EG-VEGF/prokineticin-1 Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human EG-VEGF (Met 1-Phe105) (P58294) was expressed, with a C-terminal polyhistidine tag.