Human Recombinant DR6/TNFRSF21 Protein (from HEK293 Cells)
Supplier: Sino Biological
10175-HCCH-100 10175-HCCH-200
103623-924EA 320.35 USD
103623-924 103623-922
Human Recombinant DR6/TNFRSF21 Protein (from HEK293 Cells)
Proteins and Peptides
The mature form of human DR6 (NP_055267.1) extracellular domain (Met 1-Leu 350) with five amino acids (DDDDK) at the C-terminus was expressed and purified.