Human Recombinant IL-1RAcP/IL-1R3 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10121-H08H-100 10121-H08H-100 10121-H08H-200
103623-688EA 320.35 USD
103623-688 103623-686 CA103623-686
Human Recombinant IL-1RAcP/IL-1R3 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL1R3 (NP_002173.1) extracellular domain (Met 1-Glu 359) was expressed, fused with the a C-terminal polyhistidine tag.