Human Recombinant IL1RL1/ST2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10105-H08H-200 10105-H08H-100 10105-H08H-100
103623-828EA 199.98 USD
103623-828 CA103623-828 103623-830
Human Recombinant IL1RL1/ST2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL1RL1 isoform 2 (NP_003847.2) (Met1-Phe328) was expressed fused with a polyhistidine tag at the C-terminus.