Human Recombinant Glypican 5/GPC5 Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10079-H08B-100 10079-H08B-100
103623-796EA 300.98 USD
103623-796 CA103623-796
Human Recombinant Glypican 5/GPC5 Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) was fused with a polyhistidine tag at the C-terminus.