Human Recombinant VEGFR2/Flk-1/CD309/KDR Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10012-H02H-100 10012-H02H-100
103620-696EA 300.98 USD
103620-696 CA103620-696
Human Recombinant VEGFR2/Flk-1/CD309/KDR Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human VEGFR2 (NP_002244.1) (Met 1-Glu 764) was fused with the Fc region of human IgG1 at the C-terminus.