Cynomolgus Recombinant HER2/ErbB2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90295-C02H-100 90295-C02H-1
76632-616EA 1938 USD
76632-616 103585-582
Cynomolgus Recombinant HER2/ErbB2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus ERBB2 (Met1-Thr652) [ Rhesus ERBB2 (XP_001090430.1,Asp121Asn,Leu122Pro); similar to Macaca fascicularis ERBB2 (EHH58073.1)], was expressed with the Fc region of human IgG1 at the C-terminus.