Cynomolgus Recombinant B7-DC/PD-L2/CD273 (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90249-C02H-1 90249-C02H-100
103585-242EA 300.98 USD
103585-242 76632-632
Cynomolgus Recombinant B7-DC/PD-L2/CD273 (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus PDCD1LG2(A4GW30) (Met1-Pro219) was expressed with the Fc region of human IgG1 at the C-terminus.