Rhesus Recombinant EphA4 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90039-C02H-100
103585-174EA 300.98 USD
103585-174
Rhesus Recombinant EphA4 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus EPHA4 (NP_001247799.1) (Met1-Thr547) was expressed, fused with the Fc region of human IgG1 at the C-terminus.