Rat Recombinant PSAP/Prosaposin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80597-R08H-100
103584-932EA 351.48 USD
103584-932
Rat Recombinant PSAP/Prosaposin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat Psap (NP_037145.2) (Met1-Asn554) was expressed with a polyhistidine tag at the C-terminus.