Rat Recombinant CADM3/NECL1/IGSF4B Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80220-R08H-100
103666-498EA 300.98 USD
103666-498
Rat Recombinant CADM3/NECL1/IGSF4B Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CADM3 (Q1WIM3) extracellular domain (Met 1-His 328) was expressed, fused with a polyhistidine tag at the C-terminus.