Rat Recombinant CD62L/L-Selectin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80046-R08H-100 80046-R08H-100
103886-950EA 250.48 USD
103886-950 CA76477-350
Rat Recombinant CD62L/L-Selectin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat SELL (NP_062050.3) (Met1-Asn332) was expressed with a polyhistidine tag at the C-terminus.