Rat Recombinant CCL5/RANTES Protein (His Tag) (from Yeast)
Supplier: Sino Biological
80038-R08Y-100 80038-R08Y-20
103666-274EA 401.98 USD
103666-274 103669-272
Rat Recombinant CCL5/RANTES Protein (His Tag) (from Yeast)
Proteins and Peptides
A DNA sequence encoding the rat CCL5 (NP_038681.2) (Ser25-Ser92) was expressed with a polyhistidine tag at the C-terminus.