Rat Recombinant KIM-1/TIM1/HACVR1 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80003-R16H-100
103666-208EA 351.48 USD
103666-208
Rat Recombinant KIM-1/TIM1/HACVR1 Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of rat HAVCR1 (NP_775172.1) (Ser 18-Val 238) was fused with a polyhistidine tag at the C-terminus and the Fc region of human IgG1 at the N-terminus.