Canine Recombinant IL17RD Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
70104-D08H-100
103666-160EA 300.98 USD
103666-160
Canine Recombinant IL17RD Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine IL17RD (XP_541835.3) (Ala27-Arg299) was expressed with a C-terminal polyhistidine tag.