Canine Recombinant IL1R2/IL1RB Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
70093-D02H-50
103669-110EA 199.98 USD
103669-110
Canine Recombinant IL1R2/IL1RB Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine IL1R2 (E2RKW1) (Met1-Glu343) was expressed with the Fc region of human IgG1 at the C-terminus.