Mouse Recombinant CSF1R/MCSF Receptor/CD115 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50059-M02H-100 50059-M02H-100
103886-872EA 300.98 USD
103886-872 CA76477-272
Mouse Recombinant CSF1R/MCSF Receptor/CD115 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) (Met1-Ser511) was expressed with the Fc region of human IgG1 at the C-terminus.