Influenza A H3N2 (A/Hong Kong/1/1968) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40715-V08H-100 40715-V08H-100
103886-426EA 351.48 USD
103886-426 CA76476-596
Influenza A H3N2 (A/Hong Kong/1/1968) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hong Kong/1/1968 (H3N2)) hemagglutinin (Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.