Influenza A H3N1 (A/swine/England/704563/1995) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40684-V08H-100 40684-V08H-100
103887-412EA 351.48 USD
103887-412 CA76477-812
Influenza A H3N1 (A/swine/England/704563/1995) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/England/704563/1995 (H3N1)) hemagglutinin (AFR76416) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.