Influenza A H1N2 (A/swine/Iowa/A01049060/2010) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40674-V08H-100 40674-V08H-100
103887-396EA 351.48 USD
CA76477-796 103887-396
Influenza A H1N2 (A/swine/Iowa/A01049060/2010) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/swine/Iowa/A01049060/2010(H1N2)) hemagglutinin (AEC23333) (Met1-Ser523), termed as HA, was expressed with a polyhistidine tag at the C-terminus.