Coxsackievirus Recombinant A16 (Cox A16) (strain G-10) VP1 Protein (Fc Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40535-V02B-100
103664-324EA 351.48 USD
103664-324
Coxsackievirus Recombinant A16 (Cox A16) (strain G-10) VP1 Protein (Fc Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the VP1 of Coxsackievirus A16 (Cox A16) (strain G-10) (AAA50478.1) (Gly568-Lys862) was expressed with the Fc region of human IgG1 at the C-terminus.