Influenza A H3N2 (A/Hanoi/EL134/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40489-V08B-100
103664-248EA 351.48 USD
103664-248
Influenza A H3N2 (A/Hanoi/EL134/2008) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Hanoi/EL134/2008 (H3N2)) hemagglutinin (AIU46089.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.