Influenza A H5N6 (A/duck/Jiangxi/95/2014) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40466-V08B-100
103664-198EA 351.48 USD
103664-198
Influenza A H5N6 (A/duck/Jiangxi/95/2014) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Jiangxi/95/2014(H5N6)) hemagglutinin (translated amino acids of EPI530054) (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.