Ebola virus EBOV (Sudan ebolavirus, strain Gulu) Recombinant Nucleoprotein/NP Protein (His Tag) (from E. coli)
Supplier: Sino Biological
40444-V07E1-100
103633-348EA 351.48 USD
103633-348
Ebola virus EBOV (Sudan ebolavirus, strain Gulu) Recombinant Nucleoprotein/NP Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the Sudan ebolavirus(strain Gulu) NP (YP_138520.1) (Gly630-Asp738) was expressed with a polyhistidine tag at the N-terminus.