Influenza A H3N2 (A/California/07/2004) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40118-V08B-100
103663-766EA 351.48 USD
103663-766
Influenza A H3N2 (A/California/07/2004) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/California/07/2004(H3N2)) hemagglutinin (ABW80975.1) (Met1-Trp530) was expressed with a C-terminal polyhistidine tag.