Influenza A H7N9 (A/Shanghai/1/2013) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40104-V08H1-100
103632-848EA 351.48 USD
103632-848
Influenza A H7N9 (A/Shanghai/1/2013) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Shanghai/1/2013(H7N9)), termed as HA1, Hemagglutinin (Met1-Gly338) was expressed with a C-terminal polyhistidine tag.