Influenza A H9N2 (A/duck/Hong Kong/448/78) Recombinant Hemagglutinin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40003-V08H1-100
103632-766EA 351.48 USD
103632-766
Influenza A H9N2 (A/duck/Hong Kong/448/78) Recombinant Hemagglutinin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Hong Kong/448/78(H9N2)) hemagglutinin (AAO46079.1) (Met1-Arg338),termed as HA1, was expressed with a polyhistidine tag at the C-terminus.