Human Recombinant ATP5D Protein (His Tag) (from E. coli)
Supplier: Sino Biological
14630-H07E-100
103662-792EA 351.48 USD
103662-792
Human Recombinant ATP5D Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human ATP5D (P30049) (Ala23-Glu168) was expressed with a polyhistidine tag at the N-terminus.