Human Recombinant LCAT Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13271-H08H-100
103624-418EA 250.48 USD
103624-418
Human Recombinant LCAT Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LCAT (NP_000220.1) (Met1-Glu440) was expressed with a polyhistidine tag at the C-terminus.