Human Recombinant CEACAM5/CD66e Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11077-H02H-100 11077-H02H-100
103885-920EA 300.98 USD
103885-920 CA76476-026
Human Recombinant CEACAM5/CD66e Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CEACAM5 (P06731.3) (Met1-Ala685) was expressed with the Fc region of human IgG1 at the C-terminus.