Human Recombinant ADAM8/CD156 (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10205-H08H-100 10205-H08H-1 10205-H08H-100
76632-168EA 2258 USD
CA76478-170 76632-168 103887-770
Human Recombinant ADAM8/CD156 (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ADAM8 (AAI15405.1) (Met1-Ser653) was expressed with a polyhistidine tag at the C-terminus.