Human Recombinant ITGAV & ITGB3 Heterodimer Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
CT098-H08H-100 CT098-H08H-100
103918-796EA 401.98 USD
103918-796 CA76487-928
Human Recombinant ITGAV & ITGB3 Heterodimer Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Val992) of human ITGAV (P06756) was fused with a his tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asp718) of human ITGB3 (P05106) constructed the plasmid 2. The two plasmids were co-expressed and the human ITGAV and ITGB3 heterodimer was purified.