Rhesus Recombinant IL17A & IL17F Heterodimer Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
CT063-K08H-100
103585-730EA 351.48 USD
103585-730
Rhesus Recombinant IL17A & IL17F Heterodimer Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus IL17A (XP_001106391.1) (Met 1 - Ala 155) and the mature form of rhesus IL17F (NP_001248216.1) (Arg 31 - Gln 163) linked by a polypeptide linker was expressed with a polyhistidine tag at the C-terminus.