Cynomolgus/Rhesus Recombinant Fractalkine/CX3CL1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90316-C02H-50 90316-C02H-50
103670-634EA 199.98 USD
103670-634 CA76453-482
Cynomolgus/Rhesus Recombinant Fractalkine/CX3CL1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus / rhesus CX3CL1 (EHH60415.1) (Met1-Gly100) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus CX3CL1 sequences are identical.