Cynomolgus Recombinant TIM3/HAVCR2 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90312-C02H-1 90312-C02H-100
76631-232EA 1938 USD
76631-232 103585-642
Cynomolgus Recombinant TIM3/HAVCR2 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus TIM3 (EHH54703.1) (Met1-Arg201) was expressed with the Fc region of human IgG1 at the C-terminus.