Cynomolgus/Rhesus Recombinant CD80/B7-1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90268-C02H-100 90268-C02H-1
76631-476EA 1618 USD
76631-476 103585-542
Cynomolgus/Rhesus Recombinant CD80/B7-1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus CD80 (AAC31555.1) (Met1-Asn242) was expressed with the Fc region of human IgG1 at the C-terminus.