Cynomolgus Recombinant B7-DC/PD-L2/CD273 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90249-C08H-100 90249-C08H-1
76631-716EA 1938 USD
76631-716 103585-246
Cynomolgus Recombinant B7-DC/PD-L2/CD273 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus PDCD1LG2 (A4GW30) (Met1-Pro219) was expressed with a polyhistidine tag at the C-terminus.