Rhesus Recombinant CD36/SCARB3 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90184-C02H-100
103585-216EA 300.98 USD
103585-216
Rhesus Recombinant CD36/SCARB3 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus CD36 (NP_001028085.1) (Gly30-Asn439) was expressed with the Fc region of human IgG1 at the C-terminus.