Rhesus Recombinant IL10RA Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90125-C02H-100
103585-520EA 351.48 USD
103585-520
Rhesus Recombinant IL10RA Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus IL10RA (XP_001092736.1) (Thr9-Asn245) was expressed, fused with the Fc region of human IgG1 at the C-terminus.