Cynomolgus Recombinant IL18R1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90121-C02H-100
103585-390EA 300.98 USD
103585-390
Cynomolgus Recombinant IL18R1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IL18R1 (F7H8F9) (Met1-Lys315) was expressed with the Fc region of human IgG1 at the C-terminus.