Rat Recombinant B7-DC/PD-L2/CD273 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80454-R02H-100
103585-504EA 300.98 USD
103585-504
Rat Recombinant B7-DC/PD-L2/CD273 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat PDCD1LG2 (NP_001101052.2) (Met1-Arg219) was expressed with the Fc region of human IgG1 at the C-terminus.