Rat Recombinant CD47 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80305-R02H-100
103666-608EA 300.98 USD
103666-608
Rat Recombinant CD47 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CD47 (P97829) (Met1-Lys140) was expressed, fused with the Fc region of human IgG1 at the C-terminus.