Rat Recombinant MDHA/MDH1 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
80232-R08E-50
103668-992EA 300.98 USD
103668-992
Rat Recombinant MDHA/MDH1 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the rat MDH1 (O88989) (Met 4-Ala 334) was expressed with a polyhistidine tag at the C-terminus.