Rat Recombinant CD93 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
80207-R08H-50
103669-482EA 179.78 USD
103669-482
Rat Recombinant CD93 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat CD93 (Q9ET61) (Met1-Lys571) was expressed, fused with a polyhistidine tag at the C-terminus.