Rat Recombinant IL13RA2/IL13R Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80204-R02H-100
103666-464EA 300.98 USD
103666-464
Rat Recombinant IL13RA2/IL13R Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat IL13RA2 (Q8VHK6) (Met1-Lys336) was expressed, fused with the Fc region of human IgG1 at the C-terminus.