Rat Recombinant Fas Ligand/FASLG/CD95L Protein (His Tag) (from Yeast)
Supplier: Sino Biological
80024-R07Y-100
103666-246EA 351.48 USD
103666-246
Rat Recombinant Fas Ligand/FASLG/CD95L Protein (His Tag) (from Yeast)
Proteins and Peptides
A DNA sequence encoding the rat Faslg (NP_037040.1) (Leu104-Leu278) was expressed with a polyhistidine tag at the N-terminus.