Rat Recombinant c-MET/HGFR Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
80004-R02H-100
103666-210EA 179.78 USD
103666-210
Rat Recombinant c-MET/HGFR Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat MET (Q2IBC7) (Met1-Asn932) was expressed with the Fc region of human IgG1 at the C-terminus.