Canine Recombinant Canf4 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
70261-D08H-250 70261-D08H-250
103887-322EA 401.98 USD
103887-322 CA76477-722
Canine Recombinant Canf4 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine Canf4 (NP_001177855.1) (Met1-Glu174) was expressed with a polyhistidine tag at the C-terminus.