Canine Recombinant TrkA/NTRK1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
70101-D01H-100
103666-154EA 300.98 USD
103666-154
Canine Recombinant TrkA/NTRK1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine NTRK1 (XP_851619.3) (Pro285-Lys410) was expressed with the Fc region of human IgG1 at the N-terminus.